missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH10/UBE2C Antibody (SAIC-41A-1) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20107JF585
This item is not returnable.
View return policy
Description
UbcH10/UBE2C Monoclonal antibody specifically detects UbcH10/UBE2C in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Mass Spectrometry,Immunocytochemistry/ Immunofluorescence
Specifications
| UbcH10/UBE2C | |
| Monoclonal | |
| Janelia Fluor 585 | |
| cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C | |
| Peptide 'GISAFPESDNLFK', derived from the Ubiquitin-conjugating enzyme E2C, conjugated to KLH. | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators | |
| 11065 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, ELISA, Western Blot, Mass Spectrometry, Immunocytochemistry/Immunofluorescence | |
| SAIC-41A-1 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction