missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBC9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517043
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| UBC9 | |
| Polyclonal | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P63279, P63280, P63281 | |
| UBE2I | |
| Synthetic peptide corresponding to residues surrounding V148 of human Ubc9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| UBE2I | |
| 5830467E05Rik; C358B7.1; F830028O17Rik; mUBC9; OTTHUMP00000198131; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; UBC9; UbcE2A; Ubce2i; Ubce9; Ube2i; Ubiquitin carrier protein 9; ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2 I; ubiquitin conjugating enzyme E2I; Ubiquitin conjugating enzyme E2I (homologous to yeast UBC9); Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; ubiquitin-protein ligase I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22196, 25573, 7329 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction