missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBC3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566358
This item is not returnable.
View return policy
Description
Immunogen sequence: IKTKVPSNDNS SDLLYDDLYD DD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Protein kinase CK2 is a ubiquitous and pleiotropic Ser/Thr protein kinase involved in cell growth and transformation. This gene encodes a protein similar to the E2 ubiquitin conjugating enzyme UBC3/CDC34. Studies suggest that CK2-dependent phosphorylation of this ubiquitin-conjugating enzyme functions by regulating beta-TrCP substrate recognition and induces its interaction with beta-TrCP, enhancing beta-catenin degradation.
Specifications
| UBC3B | |
| Polyclonal | |
| Unconjugated | |
| UBE2R2 | |
| 1200003M11Rik; CDC34B; cell division cycle 34 homolog; E2 ubiquitin-conjugating enzyme R2; E2-CDC34B; LOC689226; similar to ubiquitin-conjugating enzyme E2R 2; UBC3B; Ube2r2; Ubiquitin carrier protein R2; ubiquitin conjugating enzyme E2 R2; ubiquitin conjugating enzyme E2R 2; ubiquitin-conjugating enzyme E2 R2; ubiquitin-conjugating enzyme E2-32k; ubiquitin-conjugating enzyme E2-CDC34B; ubiquitin-conjugating enzyme E2R 2; ubiquitin-conjugating enzyme UBC3B; ubiquitin-protein ligase R2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q712K3 | |
| UBE2R2 | |
| Recombinant Human UBC3B. Recombinant protein control fragment (Product #RP-105163). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction