missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Anti-Ubc13 is a monoclonal antibody specific to the approximately19 kDa Ubc13 protein. Ubc13 is a ubiquitin conjugating enzyme that is complexed with a ubiquitin activating enzyme (E1), Mms2. The association of Mms2 to Ubc13 forms an active heterodimer that is involved in error-free post replicative DNA repair in yeast. Upon complexing with another conjugating enzyme variant, Uev1a, Ubc13 participates in the regulation of NF-kB signal transduction pathway in humans.
Specifications
Specifications
| Antigen | UBC13 |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.22 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | UBE2N |
| Gene Accession No. | P61088, P61089, Q9EQX9 |
| Gene Alias | BLU, UBC13, UbcH ben, UBE2N, Ubiquitin carrier protein N, Ubiquitin protein ligase N |
| Gene Symbols | UBE2N |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?