missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBC13 Polyclonal Antibody

Description
A suggested positive control is human small intestine tissue lysate. PA5-20080 can be used with blocking peptide PEP-0198.
The UBE2V1-UBE2N and UBE2V2-UBE2N heterodimers catalyze the synthesis of non-canonical 'Lys-63'-linked polyubiquitin chains. This type of polyubiquitination does not lead to protein degradation by the proteasome. Mediates transcriptional activation of target genes. Plays a role in the control of progress through the cell cycle and differentiation. Plays a role in the error-free DNA repair pathway and contributes to the survival of cells after DNA damage. Acts together with the E3 ligases, HLTF and SHPRH, in the 'Lys-63'-linked poly-ubiquitination of PCNA upon genotoxic stress, which is required for DNA repair. Appears to act together with E3 ligase RNF5 in the 'Lys-63'-linked polyubiquitination of JKAMP thereby regulating JKAMP function by decreasing its association with components of the proteasome and ERAD. Promotes TRIM5 capsid-specific restriction activity and the UBE2V1-UBE2N heterodimer acts in concert with TRIM5 to generate 'Lys-63'-linked polyubiquitin chains which activate the MAP3K7/TAK1 complex which in turn results in the induction and expression of NF-kappa-B and MAPK-responsive inflammatory genes. Together with RNF135 and UB2V1, catalyzes the viral RNA-dependent 'Lys-63'-linked polyubiquitination of RIG-I/DDX58 to activate the downstream signaling pathway that leads to interferon beta production. UBE2V1-UBE2N together with TRAF3IP2 E3 ubiquitin ligase mediate 'Lys-63'-linked polyubiquitination of TRAF6, a component of IL17A-mediated signaling pathway. (Uniprot).
Specifications
Specifications
| Antigen | UBC13 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | UBE2N |
| Gene Accession No. | P61088, P61089, Q9EQX9 |
| Gene Alias | 1500026J17Rik; AL022654; BB101821; bendless protein; bendless-like ubiquitin conjugating enzyme; bendless-like ubiquitin-conjugating enzyme; Blu; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; HEL-S-71; MGC131857; MGC8489; UBC13; UbcH13; UbcH-ben; UBCHBEN; UBC13; UBE2N; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2 N; ubiquitin conjugating enzyme E2N; ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); ubiquitin-protein ligase N; yeast UBC13 homolog |
| Gene Symbols | UBE2N |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?