Learn More
UBC12 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H8D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67482-100UL
Description
The UBC12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBC12. This antibody recognizes human, mouse, rat antigen. The UBC12 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| UBC12 | |
| 1H8D2 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| hUbc12, NEDD8 carrier protein, NEDD8 conjugating enzyme Ubc12, NEDD8 protein ligase, UBC RS2, UBC12, UBE2M | |
| 100 μL | |
| 9040 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |