Learn More
Paragon Scientific™ Ubbelohde Viscometer for Tranparent Liquids (UKAS ISO 17025 certified)
Product Code. 16090490
Click to view available options
Viscosity Range:
0.3 to 1 cSt
0.6 to 3 cSt
1 to 5 cSt
10 to 50 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1000 to 5000 cSt
2 to 10 cSt
20 to 100 cSt
20,000 to 100,000 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Unit Size:
Each
Product Code. 16090490
Brand: Paragon Scientific™ UBBEL13UKC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Specifications
Viscosity Range 2000 to 10,000 cSt
Calibrated Yes
Approximate Constant 10
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Includes UKAS ISO 17025 calibration
Size 4
8 week lead-time