missing translation for 'onlineSavingsMsg'
Learn More

UBASH3B/STS1/Tula-2 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30737649 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737649 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737649 Supplier Novus Biologicals Supplier No. AF6696A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

UBASH3B/STS1/Tula-2 Polyclonal antibody specifically detects UBASH3B/STS1/Tula-2 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBASH3B/STS1/Tula-2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias Cbl-interacting protein p70, Cbl-interacting protein Sts-1, KIAA1959SH3 domain-containing 70 kDa protein, suppressor of T-cell receptor signaling1, nm23-phosphorylated unknown substrate, MGC15437, nm23-phosphorylated unknown substrate, P70, STS1EC 3.1.3.48, STS-1ubiquitin-associated and SH3 domain-containing protein B, Suppressor of T-cell receptor signaling 1, T-cell ubiquitin ligand 2, TULA-2, Tyrosine-protein phosphatase STS1/TULA2, ubiquitin associated and SH3 domain containing B
Host Species Sheep
Immunogen E. coli-derived recombinant human UBASH3B/STS1, Gly48-Phe253, Accession # Q8TF42
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84959
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.