missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBASH3B, Mouse anti-Human, Clone: 3G7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00084959-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: DGVLERRPYEDQGLGETTPLTIICQPMQPLRVNSQPGPQKRCLFVCRHGERMDVVFGKYWLSQCFDAKGRYIRTNLNMPHSLPQRSGGFRDYEKDAPISpecifications
| UBASH3B | |
| Monoclonal | |
| Unconjugated | |
| ubiquitin associated and SH3 domain containing, B | |
| KIAA1959/MGC15437/STS-1/STS1/p70 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3G7 | |
| In 1x PBS, pH 7.4 | |
| NM_032873 | |
| UBASH3B | |
| UBASH3B (NP_116262.2, 344 a.a. ∼ 441 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 84959 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction