missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UBAP2L Polyclonal antibody specifically detects UBAP2L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | UBAP2L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Host Species | Rabbit |
| Immunogen | UBAP2L antibody was raised against a 19 amino acid peptide from near the amino terminus of human UBAP2L. The immunogen is located within amino acids 200 - 250 of UBAP2L. Amino Acid Squence: DDNYGNSSGNTWNNTGHFE |
| Quantity | 0.1 mL |
| Research Discipline | Cell Biology |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?