missing translation for 'onlineSavingsMsg'
Learn More

UBAP2L Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18402276 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402276 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402276 Supplier Novus Biologicals Supplier No. NBP281871AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBAP2L Polyclonal antibody specifically detects UBAP2L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBAP2L
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias FLJ42300, KIAA0144NICE-4, NICE4, Protein NICE-4, ubiquitin associated protein 2-like, ubiquitin-associated protein 2-like
Host Species Rabbit
Immunogen UBAP2L antibody was raised against a 19 amino acid peptide from near the amino terminus of human UBAP2L. The immunogen is located within amino acids 200 - 250 of UBAP2L. Amino Acid Squence: DDNYGNSSGNTWNNTGHFE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9898
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.