missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552174
This item is not returnable.
View return policy
Description
Immunogen sequence: MEMGFDEKEV IDALRVNNNQ QNAACEWLLG DRKPSPEELD KGIDPDSPLF QAILDNPVVQ LGLTNPKTLL AFEDMLENPL NSTQWMNDPE TGPVMLQISR IFQTLNRT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.
Specifications
| UBAC1 | |
| Polyclonal | |
| Unconjugated | |
| UBAC1 | |
| 1110033G07Rik; AA407978; E3 ubiquitin-protein ligase subunit KPC2; GBDR1; Gdbr1; glialblastoma cell differentiation-related protein 1; Kip1 ubiquitination-promoting complex protein 2; KPC2; putative glialblastoma cell differentiation-related; putative glialblastoma cell differentiation-related protein; RP11-432J22.3; testicular secretory protein Li 64; UBA domain containing 1; UBA domain-containing protein 1; Ubac1; Ubadc1; ubiquitin associated domain containing 1; ubiquitin-associated domain-containing protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10422, 362087, 98766 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XIR9, Q8VDI7, Q9BSL1 | |
| UBAC1 | |
| Recombinant protein corresponding to Human UBAC1. Recombinant protein control fragment (Product #RP-94774). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction