Learn More
UBAC1 Mouse anti-Human, Unconjugated, Clone: 2A3G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67385-1-IG-20UL
This item is not returnable.
View return policy
Description
The UBAC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBAC1. This antibody recognizes human, mouse, rat antigen. The UBAC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UBAC1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 2A3G7 | |
| Unconjugated | |
| 20 μL | |
| 10422 | |
| IgG1 |