missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88530
This item is not returnable.
View return policy
Description
UBAC1 Polyclonal specifically detects UBAC1 in Human samples. It is validated for Western Blot.
Specifications
| UBAC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10422 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| E3 ubiquitin-protein ligase subunit KPC2, GBDR1UBA domain-containing protein 1, Glialblastoma cell differentiation-related protein 1, Kip1 ubiquitination-promoting complex protein 2, KPC2, putative glialblastoma cell differentiation-related protein, RP11-432J22.3, UBA domain containing 1, UBADC1, ubiquitin associated domain containing 1, ubiquitin-associated domain-containing protein 1 | |
| The immunogen is a synthetic peptide directed towards the middle region of human UBAC1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction