missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cell Biology |
| Target | UBAC1 |
| Synonym | E3 ubiquitin-protein ligase subunit KPC2, GBDR1UBA domain-containing protein 1, Glialblastoma cell differentiation-related protein 1, Kip1 ubiquitination-promoting complex protein 2, KPC2, putative glialblastoma cell differentiation-related protein, RP11-432J22.3, UBA domain containing 1, UBADC1, ubiquitin associated domain containing 1, ubiquitin-associated domain-containing protein 1 |
| Gene Symbol | UBAC1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?