Learn More
Invitrogen™ UBA5 Polyclonal Antibody

Description
Immunogen sequence: MAESVERLQQ RVQELERELA QERSLQVPRS GDGGGGRVRI EKMSSEVVDS NPYSRLMALK RMGIVSDYEK IRTFAVAIVG VGGVGSVTAE MLTRCGIGKL LLFDYDKVEL ANMNRLFFQP HQAGLSKVQA AEHTLRNINP DVLFEVHNYN ITTVENFQHF MDRISNGGLE EGKPVDLVLS CVDNFEARMT INTACNELGQ TWMESGVSEN AVSGHIQLII PGESACFACA PPLVVAANID EKTLKREGVC AASLPTTMGV VAGILVQNVL KFLLNFGTVS FYLGYNAMQD FFPTMSMKPN PQCDDRNCRK QQEEYKKKVA ALPKQEVIQE EEEIIHEDNE WGIELVSEVS EEELKNFSGP VPDLPEGITV AYTIPKKQED SVTELTVEDS GESLEDLMAK MKNM; Positive Samples: HT-29, A-549, Jurkat, HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus.
Specifications
Specifications
| Antigen | UBA5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | UBA5 |
| Gene Accession No. | Q5M7A4, Q8VE47, Q9GZZ9 |
| Gene Alias | 5730525G14Rik; AW240750; cb972; FLJ23251; FLJ23251UBA5; THIFP1; UBA5; UBA5, ubiquitin-activating enzyme E1 homolog; ube1dc1; ubiquitin like modifier activating enzyme 5; ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; ubiquitin-like modifier-activating enzyme 5; ubiquitin-like modifier-activating enzyme 5-like protein; UFM1-activating enzyme |
| Gene Symbols | UBA5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.