Learn More
UBA5 Mouse, anti-Human, Clone: 3E5C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68537-1-IG-20UL
This item is not returnable.
View return policy
Description
The UBA5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBA5. This antibody recognizes human, mouse, rat antigen. The UBA5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UBA5 | |
| 3E5C8 | |
| FLJ17281, FLJ23251, FLJ23251UBA5, THIFP1, UBA5, UBE1DC1, Ubiquitin activating enzyme 5, UFM1 activating enzyme | |
| 20 μL | |
| 79876 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG1 |