missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | UBA3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228797
|
Novus Biologicals
NBP3-33221-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232880
|
Novus Biologicals
NBP3-33221-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBA3 Monoclonal antibody specifically detects UBA3 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| UBA3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Cycle and Replication, Neuroscience | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 9039 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp566J164, EC 6.3.2, EC 6.3.2.-, hUba3, MGC22384, NEDD8-activating enzyme E1 catalytic subunit, NEDD8-activating enzyme E1C, Nedd8-activating enzyme hUba3, UBA3, ubiquitin-activating enzyme E1 homolog, UBE1C, Ubiquitin-activating enzyme 3, Ubiquitin-activating enzyme E1C, ubiquitin-activating enzyme E1C (homologous to yeast UBA3), ubiquitin-like modifier activating enzyme 3, Ubiquitin-like modifier-activating enzyme 3, yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-463 of human UBA3 (NP_003959.3).,, Sequence:, MADGEEPEKKRRRIEELLAEKMAVDGGCGDTGDWEGRWNHVKKFLERSGPFTHPDFEPSTESLQFLLDTCKVLVIGAGGLGCELLKNLALSGFRQIHVIDMDTIDVSNLNRQFLFRPKDIGRPKAEVAAEFLNDRVPNCNVVPHFNKIQDFNDTFYRQFHIIVCGLDSIIARRWINGMLISLLNYEDGVLDPSSIVPLIDGGTEGFKGNARVILPGMTACIECTLELYPPQVNFPMCTIASMPRLPEHCIEYVRMLQWPKEQPFGEGVPLDGDDPEHIQWIFQKSLERASQYNIRGVTYRLTQGVVKRIIPAVASTNAVIAAVCATEVFKIATSAYIPLNNYLVFNDVDGLYTYTFEAERKENCPACSQLPQNIQFSPSAKLQEVLDYLTNSASLQMKSPAITATLEGKNRTLYLQSVTSIEERTRPNLSKTLKELGLVDGQELAVADVTTPQTVLFKLHFTS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title