missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA3 Antibody (1B6-6-5-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-33073
This item is not returnable.
View return policy
Description
UBA3 Monoclonal antibody specifically detects UBA3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| UBA3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| DKFZp566J164, EC 6.3.2, EC 6.3.2.-, hUba3, MGC22384, NEDD8-activating enzyme E1 catalytic subunit, NEDD8-activating enzyme E1C, Nedd8-activating enzyme hUba3, UBA3, ubiquitin-activating enzyme E1 homolog, UBE1C, Ubiquitin-activating enzyme 3, Ubiquitin-activating enzyme E1C, ubiquitin-activating enzyme E1C (homologous to yeast UBA3), ubiquitin-like modifier activating enzyme 3, Ubiquitin-like modifier-activating enzyme 3, yeast) | |
| Recombinant protein within Human UBA3 aa 220-424/ 463. (Uniprot: Q8TBC4) | |
| 100 μL | |
| Apoptosis, Cell Cycle and Replication, Neuroscience | |
| 9039 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1B6-6-5-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction