missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3042800135337

Additional Details : Weight : 187.16040kg

 View more versions of this product

Product Code. 15944456

  • £2535.52 / Each
Estimated Shipment: 05-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly:  drawers remain open in any position, interior of drawers completely visible – access to all containers
  • Safe: drawers fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: drawers lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Specifications

Specifications

502 mm
Safety storage under bench cabinet
979 mm
Powder-coated plain
1102 mm
183.50 kg
Light Gray
600 mm
UB90.060.110.050.2S
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
2 x drawer (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
382 mm
Steel
502 x 1102 x 600 mm
382 x 979 x 520 mm
502 mm
Sliding
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers