missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ub-K63 Recombinant Rabbit Monoclonal Antibody (JM09-67)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA532573

Product Code. 16335984

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Ubiquitin is a conserved 76 amino acid polypeptide and can affect proteasomal degradation of the protein it is bound to, or mediate interactions with other proteins related to post-translational modifications. The degradation of cellular regulatory proteins by the Uubiquitin pathway is important as it controls the cellular growth and proliferation. Ubiquitin-dependent proteolysis occurs after a covalent attachment of the peptide to a lysine residue of a protein, which involves three enzymatic reactions: E1, E2 and E3. The first reaction involves ubiquitin-activating enzyme. The third reaction uses enzyme ubiquitin ligase (E3) to transfer the activated ubiquitin from E2 to a lysine residue on a protein, or directly transfers the ubiquitin from E2 to the substrate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ub-K63
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
P0CG47, P0CG48, P0CG49, P0CG50, P0CG51, P62979, P62982, P62983, P62984, P62986, P62987, Q63429
RPS27A, UBA52, UBB, UBC
Synthetic peptide within Human K63-linkage Specific Ubiquitin aa 36-85.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
JM09-67
Unconjugated
UBB
0610006J14Rik; 2700054O04Rik; 40S ribosomal protein S27a; 60S ribosomal protein L40; AI194771; AL033289; CEP52; CEP80; D8Ertd21e; epididymis luminal protein 112; epididymis secretory protein Li 50; Gm1863; HEL112; HEL-S-50; HMG20; HUBCEP52; L40; Large ribosomal subunit protein eL40; LOC100912032; polyubiquitin B; polyubiquitin C; polyubiquitin-B; Polyubiquitin-C; ribosomal protein S27A; RPL40; Rps27a; S27A; Small ribosomal subunit protein eS31; TI-225; Uba52; UBA80; Ubb; Ubb2; UBC; UBCEP1; UBCEP2; UBCEP80; Ubiquitin; Ubiquitin A-52 residue ribosomal protein fusion product 1; ubiquitin and ribosomal protein S27a; ubiquitin B; ubiquitin C; ubiquitin carboxyl extension protein 52; ubiquitin carboxyl extension protein 80; ubiquitin/60S ribosomal fusion protein; Ubiquitin-40S ribosomal protein S27a; ubiquitin-40S ribosomal protein S27a-like; ubiquitin-52 amino acid fusion protein; ubiquitin-60S ribosomal protein L40; ubiquitin-CEP52; ubiquitin-CEP80; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2
Rabbit
Protein A
RUO
100912032, 192255, 22186, 22187, 22190, 50522, 6233, 64156, 7311, 7314, 7316, 78294
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.