missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ub-K63 Monoclonal antibody specifically detects Ub-K63 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Ub-K63 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | HWA4C4 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody is Ub-K63. |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?