missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAP56 Antibody (OTI2C5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74727CL1
This item is not returnable.
View return policy
Description
UAP56 Monoclonal antibody specifically detects UAP56 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| UAP56 | |
| Monoclonal | |
| CoraFluor 1 | |
| 56 kDa U2AF65-associated protein, ATP-dependent RNA helicase p47, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B, DEAD box protein UAP56, EC 3.6.4.13, HLA-B associated transcript 1, HLA-B-associated transcript 1 protein, spliceosome RNA helicase BAT1, UAP56D6S81EBAT1nuclear RNA helicase (DEAD family) | |
| Full length human recombinant protein of human BAT1(NP_542165) produced in HEK293T cell. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 7919 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI2C5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction