missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAP56 Antibody (3A2B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£453.00
Specifications
| Antigen | UAP56 |
|---|---|
| Clone | 3A2B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
UAP56 Monoclonal specifically detects UAP56 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| UAP56 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Proteases & Other Enzymes | |
| PBS with 0.05% Sodium Azide | |
| 7919 | |
| Purified recombinant fragment of human DDX39B (AA: 1-250) expressed in E. Coli. | |
| Primary | |
| 49 kDa |
| 3A2B2 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 56 kDa U2AF65-associated protein, ATP-dependent RNA helicase p47, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B, DEAD box protein UAP56, EC 3.6.4.13, HLA-B associated transcript 1, HLA-B-associated transcript 1 protein, spliceosome RNA helicase BAT1, UAP56D6S81EBAT1nuclear RNA helicase (DEAD family) | |
| DDX39B | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title