missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAP1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18444AF594
This item is not returnable.
View return policy
Description
UAP1 Polyclonal antibody specifically detects UAP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UAP1 | |
| Polyclonal | |
| Western Blot | |
| AGX, AGX1, AGX2, Antigen X, EC 2.7.7, EC 2.7.7.23, EC 2.7.7.27, SPAG2, sperm associated antigen 2, Sperm-associated antigen 2, UDP-N-acetylhexosamine pyrophosphorylase, UDP-N-acteylglucosamine pyrophosphorylase 1 | |
| Recognizes region between residues Between Between 472 and 522.. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 6675 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction