missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAF1/WDR48 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95220-0.02ml
This item is not returnable.
View return policy
Description
UAF1/WDR48 Polyclonal antibody specifically detects UAF1/WDR48 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| UAF1/WDR48 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| KIAA1449DKFZp686G1794, P80, UAF1, USP1 associated factor 1, USP1-associated factor 1, WD repeat domain 48, WD repeat endosomal protein, WD repeat-containing protein 48 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-677 of human WDR48 (NP_065890.1). FSSPDGSDPKLNLGGLLLQALLEYWPRTHVNPMDEEENEVNHVNGEQENRVQKGNGYFQVPPHTPVIFGEAGGRTLFRLLCRDSGGETESMLLNETVPQWVIDITVDKNMPKFNKIPFYLQPHASSGAKTLKKDRLSASDMLQVRKVMEHVYEKIINLDNESQTTSSSNNEKPGEQEKEEDIAVLAEEKIELLCQDQVLDPNMDLRTVKHFIWKSGGDLTLHYRQKST | |
| 0.02 mL | |
| Cell Biology | |
| 57599 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction