missing translation for 'onlineSavingsMsg'
Learn More

UACA Antibody (AE-5), DyLight 405, Novus Biologicals™

Product Code. 18393672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393672 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393672 Supplier Novus Biologicals Supplier No. NBP311314V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UACA Monoclonal antibody specifically detects UACA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UACA
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone AE-5
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats
Host Species Mouse
Immunogen Nuclei of myeloid leukemia biopsy cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55075
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.