missing translation for 'onlineSavingsMsg'
Learn More

U2AF2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18432513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432513 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432513 Supplier Novus Biologicals Supplier No. NBP204139AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

U2AF2 Polyclonal antibody specifically detects U2AF2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen U2AF2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 380 and 430 of human U2 Small Nuclear Ribonucleoprotein Auxiliary Factor (65kD) using the numbering given in entry NP_009210.1 (GeneID 11338).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11338
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.