missing translation for 'onlineSavingsMsg'
Learn More

U2AF1L4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18308656 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18308656 20 μL 20µL
18351726 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18308656 Supplier Novus Biologicals Supplier No. NBP30451220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

U2AF1L4 Polyclonal specifically detects U2AF1L4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen U2AF1L4
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ35525, MGC33901, splicing factor U2AF 26 kDa subunit, U2 auxiliary factor 26, U2 small nuclear RNA auxiliary factor 1-like 3, U2 small nuclear RNA auxiliary factor 1-like 4, U2 small nuclear RNA auxiliary factor 1-like protein 3, U2 small nuclear RNA auxiliary factor 1-like protein 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 3, U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3, U2AF1L3, U2AF1L3V1, U2AF1-like protein 3, U2AF1RS3, U2AF1-RS3, U2af26
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-202 of human U2AF1L4 (NP_659424.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199746
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.