missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF1L4, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00199746-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MAEYLASIFGTEKDKVNCSFYFKIGVCRHGDRCSRLHNKPTFSQEVFTELQEKYGEIEEMNVCDNLGDHVVGNVYVKFRREEDGERAVAELSNRWFNGQAVHGNVPEVASATSCICGPFPRTSRGSSMGGDPGAGHPRGSILATIPERGTIGVPLITGMAASEALAPLPFTPNRDRCSWQDLSSKPPSLSCPILPRLPGSIMSpecifications
| U2AF1L4 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC021186.1 | |
| U2AF1L4 | |
| U2AF1L4 (AAH21186.1, 1 a.a. ∼ 202 a.a) full-length human protein. | |
| RUO | |
| 199746 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| U2 small nuclear RNA auxiliary factor 1-like 4 | |
| FLJ35525/MGC33901/U2AF1-RS3/U2AF1L3/U2AF1L3V1/U2AF1RS3/U2af26 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction