missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ U2AF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528510
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (94%), Chicken (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to the splicing factor SR family of genes. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| U2AF1 | |
| Polyclonal | |
| Unconjugated | |
| U2AF1 | |
| 2010107D16Rik; 35kDa; FP793; RN; RNU2AF1; Splicing factor U2AF 35 kDa subunit; Splicing factor U2AF 35 kDa subunit-like protein; splicing factor U2AF 35kDa subunit; U2 auxiliary factor 35 kDa subunit; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF) 1; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF),35 kDa; U2 small nuclear ribonucleoprotein auxillary factor, 35-KD subunit; U2 small nuclear RNA auxiliary factor 1; U2 small nuclear RNA auxiliary factor 1-like protein 5; U2 small nuclear RNA auxillary factor 1; U2 snRNP auxiliary factor small subunit; U2(RNU2) small nuclear RNA auxiliary factor binding protein; U2af1; U2AF1L5; U2AF35; U2AFBP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108121, 7307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.34 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q01081, Q9D883 | |
| U2AF1 | |
| Recombinant protein encompassing a sequence within the center region of human U2AF35. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction