missing translation for 'onlineSavingsMsg'
Learn More

U18666A, MedChemExpress

Product Code. 16494128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494128 10 mg 10mg
16406837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16494128 Supplier Medchem Express Supplier No. HY107433
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress U18666A, an intra-cellular cholesterol transport inhibitor, inhibits replication of Ebola virus, dengue virus, and human hepatitis C virus.

U18666A, an intra-cellular cholesterol transport inhibitor, inhibits replication of Ebola virus, dengue virus, and human hepatitis C virus.

Specifications

Chemical Name or Material U18666A
Physical Form Powder
CAS 3039-71-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H42ClNO2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 50 mg/mL (117.91 mM; Need ultrasonic) ∣H2O : 5.56 mg/mL (13.11 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]12[C@]3([H])[C@](CC=C1C[C@@H](OCCN(CC)CC)CC2)([H])[C@@]4([H])[C@@](C)(C(CC4)=O)CC3.Cl
Molecular Weight (g/mol) 424.06
Formula Weight 424.06
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.