missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U11/U12-35K Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93775-0.02ml
This item is not returnable.
View return policy
Description
U11/U12-35K Polyclonal antibody specifically detects U11/U12-35K in Human, Mouse samples. It is validated for Western Blot
Specifications
| U11/U12-35K | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HM1, HM-1, MGC138160, Protein HM-1, small nuclear ribonucleoprotein 35kDa (U11/U12), U1 snRNP-binding protein homolog, U11/U12 small nuclear ribonucleoprotein 35 kDa protein, U11/U12 snRNP 35 kDa protein, U11/U12 snRNP 35K, U11/U12-35K, U1-snRNP binding protein homolog, U1SNRNPBPU1 snRNP binding protein homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human SNRNP35 (NP_073208.1). MNDWMPIAKEYDPLKAGSIDGTDEDPHDRAVWRAMLARYVPNKGVIGDPLLTLFVARLNLQTKEDKLKEVFSRYGDIRRLRLVRDLVTGFSKGYAFIEYKEERAVIKAYRDADGLVIDQHEIFVDYELERTLKGWIPRRLGGGLGGKKES | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11066 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction