missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ u-Plasminogen Activator/Urokinase Antibody Pair [HRP]

Product Code. 18245587 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18245587

Brand: Novus Biologicals™ NBP27930015PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human u-Plasminogen Activator/Urokinase.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 4.69 pg/mL
Assay Sensitivity 4.69-300 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human u-Plasminogen Activator/Urokinase.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator
Gene ID (Entrez) 5328
Gene Symbol PLAU
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 15 ng of recombinant Human u-Plasminogen Activator/Urokinase.
  • Capture Antibody: 1 mg/mL of mouse anti-Human u-Plasminogen Activator/Urokinase monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human u-Plasminogen Activator/Urokinase monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target u-Plasminogen Activator/Urokinase
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 300 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.