Learn More
uvex U-Fit Lite Nitrile Chemical Protection Gloves
Product Code. 15317152 Shop All Uvex Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15317152 7 Each
15397142 9 Each
15327152 10 Each
15307152 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15317152 Supplier uvex Supplier No. 6059707
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Uvex U-Fit Lite disposable safety gloves are extremly lightweight, while still providing good protection against dirt and oil. Gloves are touchscreen compatible and ideal for light precision assembly work.
  • Lightweight, durable disposable NBR gloves
  • Texturized fingertips for enhanced grip
  • Reliable spray protection when handling chemicals such as acids, alkalis, solids, or aqueous saline solutions
  • Silicone-free according to the imprint test
  • Touchscreen compatible
  • Food safe
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Disposable Single-use
Exterior Finish Smooth
Latex Free Yes
Material Nitrile Rubber
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Product Type Nitrile Gloves
Size 7
Sterility Non-sterile
EN ISO 374-1 Type C
EN ISO 374-1 CHEMICAL LETTER CODES K
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Quantity Box of 100