Learn More
uvex U-Fit Disposable NBR Protective Gloves
Product Code. 15357142 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
Product Code. 15357142
Brand: uvex 6059608
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thin, reliable NBR gloves feature a roughed glove surface for enhanced grip capabilities and provide reliable splash protection when handling chemicals such as acids, alkalis, solids, and aqueous salt solutions.
  • Thin, reliable NBR gloves enhanced grip capabilities due to the roughened palm surface
  • Splash protection when handling chemicals such as acids, alkalis, solids, or aqueous salt solutions
  • Silicone-free according to the impression test
  • Highly flexible with good tactile sensitivity
  • Very good mechanical strength
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Latex Free Yes
Material NBR
Palm Thickness (Metric) 0.1 mm
Product Type Chemical-resistant Gloves
Size 8
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KPT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection