Learn More
Mettler Toledo™ U-Electrode, Universal
Shop All Mettler Toledo Products
Click to view available options
:
Size: Large
Description
- Large electrode for discharging all kinds of samples and tare containers with dimensions of 15x25cm
- Improves weighing accuracy
- Enables fast dosing
- Reduces contamination risk
- Can be freely placed next to your balance
Specifications
Specifications
| For Use With (Equipment) | Laboratory Balance |
| Electrical Requirements | 100 to 240 V 50/60 Hz |
| For Use With (Application) | Discharging electrostatic charge |
| Size | Large |