missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ U-Chem 3200 Chemical Protection Gloves

Product Code. 17260531 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17260531

Brand: Uvex™ 6097211

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

NBR-coated chemical protection gloves feature a slim-fit design to provide high levels of dexterity. These gloves are ideal for handling oil, cooling lubricants, and chemicals.

  • NBR-coated chemical protection gloves with nylon weave supporting material
  • Slim-fit design easily stretches and naturally moves with the hand for increased dexterity
  • Additional coating on the palm ensures a secure grip
  • Resistant to grease, mineral oils, and various chemicals
  • Ideal for handling oil, cooling lubricants, and grease
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Blue, Black
Size 11
EN388 Standard 3131X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLMOT
For Use With (Application) Handling Oil, Cooling Lubricants, and Chemicals
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Nylon
Series U-Chem 3200
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.