missing translation for 'onlineSavingsMsg'
Learn More

U-73343, MedChemExpress

Product Code. 16426937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16426937 Supplier Medchem Express Supplier No. HY108630
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress U-73343, works as a protonophore, is an inactive analog of U-73122 and can be used as a negative control. U-73343 dose-dependently inhibits acid secretion irrespective of the stimulant. U-73122 is a phospholipase C (PLC) and 5-LO (5-lipoxygenase) inhibitor with an IC50 of 1-2.1 μM for PLC.

U-73343, works as a protonophore, is an inactive analog of U-73122 and can be used as a negative control. U-73343 dose-dependently inhibits acid secretion irrespective of the stimulant. U-73122 is a phospholipase C (PLC) and 5-LO (5-lipoxygenase) inhibitor with an IC50 of 1-2.1 μM for PLC.

Specifications

Chemical Name or Material U-73343
CAS 142878-12-4
Color White
Purity Grade Notes Research
Molecular Formula C29H42N2O3
Quantity 5 mg
Solubility Information DMSO : 5 mg/mL (10.71 mM; Need ultrasonic)
SMILES O=C(CC1)N(CCCCCCN[C@H]2CCC3C4CCC5=C(C=CC(OC)=C5)C4CC[C@]23C)C1=O
Molecular Weight (g/mol) 466.66
Formula Weight 466.66
Percent Purity 99.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.