missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U-2 OS (human osterogenic sarcoma) Whole cell lysate, Non-denatured; Abnova
Description
- Tissue: Bone
- Host: Human
- Lysis Buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting, Immunoprecipitation
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Bone |
| Description | Whole cell lysate (non-denatured) |
| Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 200 μg |
| Storage Buffer | In modified RIPA Lysis Buffer. |
| Host Species | Human |
| Content And Storage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Safety and Handling
Product Identifier
- 200UG U-2 OS (human osterogenic sarcoma) whole cel
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P102-Keep out of reach of children.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: potassium phosphatemonobasic, sodium phosphate dibasic
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?