missing translation for 'onlineSavingsMsg'
Learn More

U-104, MedChemExpress

Product Code. 16485718
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16485718

Brand: Medchem Express HY13513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress U-104 (SLC-0111) is a potent carbonic anhydrase (CA) inhibitor for CA IX and CA XII with Ki values of 45.1 nM and 4.5 nM, respectively. U-104 shows a significant delay in tumor growth in mice model.

U-104 (SLC-0111) is a potent carbonic anhydrase (CA) inhibitor for CA IX and CA XII with Ki values of 45.1 nM and 4.5 nM, respectively. U-104 shows a significant delay in tumor growth in mice model.

Specifications

Chemical Name or Material U-104
CAS 178606-66-1
Color White
Purity Grade Notes Research
Molecular Formula C13H12FN3O3S
Quantity 10 mg
Synonym SLC-0111
Solubility Information DMSO : ≥ 100 mg/mL (323.29 mM)
SMILES O=S(C1=CC=C(NC(NC2=CC=C(F)C=C2)=O)C=C1)(N)=O
Molecular Weight (g/mol) 309.32
Formula Weight 309.32
Percent Purity 99.73%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.