missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TYW1 Recombinant Protein Antigen

Product Code. 18293894 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18293894 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18293894 Supplier Novus Biologicals™ Supplier No. NBP258222PEP
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TYW1. Source: E.coli Amino Acid Sequence: GFATVLAEAVTSLDLPVAIINLKEYDPDDHLIEEVTSKNVCVFLVATYTDGLPTESAEWFCKWLEEASIDFRFGKTYLKGMR The TYW1 Recombinant Protein Antigen is derived from E. coli. The TYW1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55253
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name TYW1 Recombinant Protein Antigen
Content And Storage Store at −20C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias FLJ10900, MGC23001, MGC60291, Radical S-adenosyl methionine and flavodoxin domain-containing protein 1, radical S-adenosyl methionine and flavodoxin domains 1, RSAFD1, tRNA wybutosine-synthesizing protein 1 homolog, tRNA-yW synthesizing protein 1 homolog
Gene Symbol TYW1
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50963. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.