Learn More
DuPont™ Sterile Tyvek™ IsoClean Overboots
Product Code. 15483123
Click to view available options
Unit Size:
Pack of 100
Product Code. 15483123
Brand: DuPont™ D14246800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use overboots in cleanroom class ISO 5 and higher environments to maintain quality. Sterile Tyvek™ IsoClean Overboots have a reinforced seam and repel water-based liquids and aerosols. Clean laundered and gamma-sterilised.
Tyvek™ IsoClean
  • Sterile overboot in white
  • Fixation ties
  • Tunnelled elastics at ankles and gripper slip-retardant full sole
  • Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Offers an excellent barrier against fine particles and fibers (down to micron in size) and is ultra-low-linting
  • Silicon non-added
  • Garments and accessories made of Tyvek™ IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments
Disposable Overboots
  • This accessory is not CE certified as chemical protective clothing
  • Clean laundered and gamma-sterilised
  • Suitable for use in cleanroom class ISO 5 and higher
  • Sterility assurance level (SAL) of 10-6
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
  • Full traceability on all sterilized apparel with certificates of sterility available
TRUSTED_SUSTAINABILITY
Specifications
Material Tyvek
Color White
Disposable Single-use
Sole Type Slip-resistant
Seam Type Reinforced seam protection
Packaging Quantity 100