Learn More
DuPont™ Tyvek™ IsoClean™ Slip-retardant Boot Covers
Composed of flash spun high-density polyethylene. DuPont™ Tyvek™ IsoClean™ Boot Covers are permeable to both air and water vapour, yet repel water-based liquids and aerosols. They offer an excellent barrier against fine particles and fibres (down to 1μ in size) and are ultra-low-linting.
Brand:  DuPont™ D14246831
Product Code. 15551975
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Fixation ties and slip-retardant sole
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
  • Single packed
  • Note: this product is not CE certified as chemical protective clothing
  • Sterile model available