Learn More
DuPont™ Tyvek™ IsoClean™ Sleeves
Product Code. 15571975 Shop All DuPont Products
Change view
Click to view available options
:
Size: Universal
Product Code.
15571975 Size: Universal
1 options
Product Code. 15571975 Supplier DuPont™ Supplier No. D14246894
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ IsoClean™ Gown is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibers down to 1μ in size.
  • White, universal size.
  • Tunnelled elastics
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
• Note: this product is not CE certified as chemical protective clothing
TRUSTED_SUSTAINABILITY
Specifications
Size Standard
Construction Flash-spun HDPE
Product Type Controlled Environment Sleeves
Material HDPE
Color White
Sterility Non-sterile
Seam Type Bound
Cuff Style Elastic
Model IC501B
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing and Electronics Industries, Other Critical or Controlled Environments
Certifications/Compliance PPE Category 1
Length (Metric) 45 cm