missing translation for 'onlineSavingsMsg'
Learn More

KERN and SOHN Protective Dust Cover

Product Code. 17836866
Encompass_Preferred
Click to view available options
Dimensions (L x W):
485 x 450 mm
650 x 750 mm
This item is not returnable. View return policy

Product Code. 17836866

Brand: KERN and SOHN OBBA1387

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For microscopes

  • Microscope dust covers give greater ease of use.
  • By using these, you can easily avoid the time-consuming cleaning required with routine use of your microscope.

Specifications

Type Dust Cover
Color Black
Dimensions (L x W) 485 x 450 mm
For Use With (Equipment) Microscopes
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.