Learn More
DuPont™ Tyvek™ 500 Boot Cover with Antislip
Product Code. 17602955 Shop All DuPont Products
Change view
Click to view available options
:
Model: POBA Overboot
Product Code.
17602955 Model: POBA Overboot
1 options
Product Code. 17602955 Supplier DuPont™ Supplier No. TYPOBASWH00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Knee-length overboot offerring enhanced protection when exposed to hazardous substance.
  • Elasticated top and fixation ties
  • Provides an ideal balance of protection, durability and comfort
  • Silicon non-added
  • Accessory
TRUSTED_SUSTAINABILITY
Specifications
Size One Size
Material Polyethylene
Color White
Sole Type Slip-resistant
Model POBA Overboot
Seam Type Stitched
For Use With (Application) Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/Remediation, General Maintenance/Operations, Spray Painting and General Clean-up, Amongst Many Others.
Certifications/Compliance (Europe) (EU) 2016/425, EN 14126, EN 1149-1