missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TYSND1 Polyclonal specifically detects TYSND1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | TYSND1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | MGC131934, trypsin domain containing 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TYSND1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?