missing translation for 'onlineSavingsMsg'
Learn More

TYSND1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18342727 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342727 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342727 Supplier Novus Biologicals Supplier No. NBP305923AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TYSND1 Polyclonal specifically detects TYSND1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYSND1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias MGC131934, trypsin domain containing 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TYSND1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219743
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.