missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A9, MedChemExpress

Product Code. 16442677
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442677 Supplier Medchem Express Supplier No. HY15511
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tyrphostin A9, a PDGFR inhibitor, is a potent inducer of mitochondrial fission. Tyrphostin A9 emerged as the most potent and selective of 51 tyrosine kinase inhibitors tested against the TNF-induced respiratory burst. Tyrphostin A9 has anti-influenza virus activities.

Tyrphostin A9, a PDGFR inhibitor, is a potent inducer of mitochondrial fission. Tyrphostin A9 emerged as the most potent and selective of 51 tyrosine kinase inhibitors tested against the TNF-induced respiratory burst. Tyrphostin A9 has anti-influenza virus activities.

Specifications

Chemical Name or Material Tyrphostin A9
CAS 10537-47-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H22N2O
Quantity 50 mg
Synonym Tyrphostin 9 Malonoben
Solubility Information DMSO : 50 mg/mL (177.07 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES N#C/C(C#N)=C/C1=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C1
Molecular Weight (g/mol) 282.38
Formula Weight 282.38
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.